Learn More
Sorbent Technologies Inc Silica Gel Desiccant Spheres w/100% Blue Indicator, 2-5mm 5kg
SDP
Catalog No. 501138805
Encompass_Preferred
Silica Gel Desiccant Spheres with 100% blue indicator are non-corrosive, non-toxic, and chemically inert. The desiccant spheres have particle distribution of 2-5mm and a quantity of 5kg. It is a highly porous form of silica, with an extremely large internal surface area and an affinity towards water and other polar compounds. It will not change its size or shape. Even when the silica gel is water saturated, it remains free-flowing. The spheres with blue indicator (cobalt chloride) turn pink upon reaching moisture capacity.
Catalog No. 50-113-8805 Supplier Sorbent Technologies Inc Supplier No. 231105
Silica Gel Desiccant Spheres with 100% blue indicator are non-corrosive, non-toxic, and chemically inert. The desiccant spheres have particle distribution of 2-5mm and a quantity of 5kg. It is a highly porous form of silica, with an extremely large internal surface area and an affinity towards water and other polar compounds. It will not change its size or shape. Even when the silica gel is water saturated, it remains free-flowing. The spheres with blue indicator (cobalt chloride) turn pink upon reaching moisture capacity.