Learn More
Sorbent Technologies Inc Silica Gel Desiccant Spheres w/100% Yellow Indicator, 2-5mm, 1kg
SDP
Catalog No. 501138807
Encompass_Preferred
Silica Gel Desiccant Spheres with 100% yellow indicator are non-corrosive and chemically inert. The desiccant spheres have particle distribution of 2-5mm and a quantity of 1kg. It is a highly porous form of silica, with an extremely large internal surface area and an affinity towards water and other polar compounds. It will not change its size or shape. Even when the silica gel is water saturated, it remains free-flowing. The spheres with yellow indicator (phenol phthalein) turn blue/green upon reaching moisture capacity.
Catalog No. 50-113-8807 Supplier Sorbent Technologies Inc Supplier No. 232101
Silica Gel Desiccant Spheres with 100% yellow indicator are non-corrosive and chemically inert. The desiccant spheres have particle distribution of 2-5mm and a quantity of 1kg. It is a highly porous form of silica, with an extremely large internal surface area and an affinity towards water and other polar compounds. It will not change its size or shape. Even when the silica gel is water saturated, it remains free-flowing. The spheres with yellow indicator (phenol phthalein) turn blue/green upon reaching moisture capacity.