Learn More
Miles Scientific SIL F/PLASTIC SHEETS 25/BX
SDP
Catalog No. NC9578016
Encompass_Preferred
Silica Gel F coated on Plastic Sheets 200 micrometer 20x20cm; 25 sheets.
Catalog No. NC9578016 Supplier Miles Scientific Supplier No. P159017
Silica Gel F coated on Plastic Sheets 200 micrometer 20x20cm; 25 sheets.