Learn More
Sorbent Technologies Inc Silica G TLC Plates, w/UV254, glass backed, 250um, 2.5x7.5cm, 100/pk
SDP
Catalog No. 501138538
Encompass_Preferred
"Silica G" is a standard grade, unmodified Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to a glass support. It has a thickness of 250um and dimensions of 2.5x7.5cm. There are 100 plates per pack.
Catalog No. 50-113-8538 Supplier Sorbent Technologies Inc Supplier No. 1615198
"Silica G" is a standard grade, unmodified Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to a glass support. It has a thickness of 250um and dimensions of 2.5x7.5cm. There are 100 plates per pack.