Learn More
Sorbent Technologies Inc Silica Gel, Premium Rf, 60A, 75-200um, 1kg
SDP
Catalog No. NC0910733
Encompass_Preferred
Silica Gel, Premium Rf has a porosity of 60A, particle distribution of 75-200um, and a quantity of 1kg. It is used as the stationary phase for classical colum chromatography where gravity flow is utilized. Premium Rf (Resolution Factor) silica gel is the highest quality granular silica gel on the market. This high-purity product provides narrow particle and pore distribution and is designed to increase yield, making it ideal for complicated separations both in the lab and in full-scale production.
Catalog No. NC0910733 Supplier Sorbent Technologies Inc Supplier No. 527001
Silica Gel, Premium Rf has a porosity of 60A, particle distribution of 75-200um, and a quantity of 1kg. It is used as the stationary phase for classical colum chromatography where gravity flow is utilized. Premium Rf (Resolution Factor) silica gel is the highest quality granular silica gel on the market. This high-purity product provides narrow particle and pore distribution and is designed to increase yield, making it ideal for complicated separations both in the lab and in full-scale production.