Learn More
Miles Scientific SIL GEL/PLASTIC SHEETS 25/BX
SDP
Catalog No. NC9644432
Encompass_Preferred
Silica Gel coated on Plastic Sheets 250micrometer 20 x 20cm 25 sheets
Catalog No. NC9644432 Supplier Miles Scientific Supplier No. P158017
Silica Gel coated on Plastic Sheets 250micrometer 20 x 20cm 25 sheets