Learn More
Grainger 3 oz Silicone Heat Sink Compound, White Paste
Catalog No. NC1833685
Encompass_Preferred
Noncuring Super Lube Silicone Heat Sink Compound is formulated with thermally conductive, fine metal oxide powders and pure silicone oil to help extend the life of heat-sensitive electronics and electrical components. It is designed to boost efficient transfer of heat away from an electrical device to its heat sink or chassis. The compound won't harden, dry out, or melt.
Catalog No. NC1833685 Supplier Grainger Supplier No. 44N787
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Noncuring Super Lube Silicone Heat Sink Compound is formulated with thermally conductive, fine metal oxide powders and pure silicone oil to help extend the life of heat-sensitive electronics and electrical components. It is designed to boost efficient transfer of heat away from an electrical device to its heat sink or chassis. The compound won't harden, dry out, or melt.