Learn More
YMC America, Inc SIL-HG guard 60A 10um 50x20mm
SDP
Catalog No. 5000881704
Encompass_Preferred
The high-purity silica found in YMC-Pack SIL columns provides virtually complete sample recovery The absence of silica impurities reduces non-specific sample adsorption and the totally porous nature of the silica provides high sample loading The elevated chemical purity of YMC silica results in a particle structure exhibiting high mechanical strength YMC-Pack SIL is suitable for separation of fat-soluble compounds using non-polar mobile phases as well as the separation of positional isomers that are difficult to separate in reversed-phase mode SIL-06 (60 pore size) has greater adsorption properties than SIL (120 pore size) due to its larger surface area and is generally useful for separating natural products with low molecular weight
Catalog No. 50-008-81704 Supplier YMC America, Inc Supplier No. SLG06S110520WTG
The high-purity silica found in YMC-Pack SIL columns provides virtually complete sample recovery The absence of silica impurities reduces non-specific sample adsorption and the totally porous nature of the silica provides high sample loading The elevated chemical purity of YMC silica results in a particle structure exhibiting high mechanical strength YMC-Pack SIL is suitable for separation of fat-soluble compounds using non-polar mobile phases as well as the separation of positional isomers that are difficult to separate in reversed-phase mode SIL-06 (60 pore size) has greater adsorption properties than SIL (120 pore size) due to its larger surface area and is generally useful for separating natural products with low molecular weight