Learn More
Sorbent Technologies Inc Resolv-Rf™ Silica Hard Layer TLC Plates, w/UV254, glass backed, 250um, 20x20cm, 25/pk
SDP
Catalog No. NC2961490
Encompass_Preferred
Resolv-Rf™ Silica Hard Layer (HL) TLC Plates are made with standard grade Silica 60, which has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical hard layer is bound to a glass support. It has a thickness of 250um and dimensions of 20x20cm. There are 25 plates per pack. Resolv-Rf™ TLC Plates are suitable for all common separations. The layer has a hardness that is abrasion resistant and can be easily labeled with a lead pencil for clear identification. The dense coating yields narrow bands for a high separation efficiency. Applications include: drug synthesis compounds, identity of drugs, food and environmental samples, quality control testing of pharmaceuticals, etc. Made in the U.S.A.
Catalog No. NC2961490 Supplier Sorbent Technologies Inc Supplier No. 9115126
Resolv-Rf™ Silica Hard Layer (HL) TLC Plates are made with standard grade Silica 60, which has a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical hard layer is bound to a glass support. It has a thickness of 250um and dimensions of 20x20cm. There are 25 plates per pack. Resolv-Rf™ TLC Plates are suitable for all common separations. The layer has a hardness that is abrasion resistant and can be easily labeled with a lead pencil for clear identification. The dense coating yields narrow bands for a high separation efficiency. Applications include: drug synthesis compounds, identity of drugs, food and environmental samples, quality control testing of pharmaceuticals, etc. Made in the U.S.A.