Learn More
Sorbent Technologies Inc Silica N HR TLC Plates, w/UV254, polyester backed, 200um, 5x20cm, 50/pk
SDP
Catalog No. 501138572
Encompass_Preferred
"Silica N HR" is a high purity Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to a plastic support. It has a thickness of 200um and dimensions of 5x20cm. There are 50 plates per pack.
Catalog No. 50-113-8572 Supplier Sorbent Technologies Inc Supplier No. 1824147
"Silica N HR" is a high purity Silica 60 with a surface area of 500 m2/g, mean pore size of 60A, specific pore volume of 0.75 ml/g, and particle size of 5-17um. The TLC plates contain UV254 fluorescent indicator. This analytical soft layer is bound to a plastic support. It has a thickness of 200um and dimensions of 5x20cm. There are 50 plates per pack.