Learn More
Accuflow Systems Saint-Gobain Versilic Stopper, Platinum-Cured, 12.5 x 16.5 x 20 mm
Catalog No. NC2327726
Encompass_Preferred
Saint-Gobain Versilic silicone stoppers are platinum-cured closures designed for laboratory and bioprocess applications. This configuration measures 12.5 mm top diameter, 16.5 mm bottom diameter, and 20 mm height, and is supplied 10 per pack.
  • Platinum-cured silicone provides high purity and broad chemical compatibility.
  • Dimensions: top 12.5 mm x bottom 16.5 mm x height 20 mm (0.493 x 0.650 x 0.788 in).
  • Supplied 10 per pack for small-batch use and inventory control.
  • Intended for closure systems in single-use assemblies and laboratory glassware.
  • Datasheet and material compliance information available from the manufacturer.
Catalog No. NC2327726 Supplier Accuflow Systems Supplier No. D1069809
Saint-Gobain Versilic silicone stoppers are platinum-cured closures designed for laboratory and bioprocess applications. This configuration measures 12.5 mm top diameter, 16.5 mm bottom diameter, and 20 mm height, and is supplied 10 per pack.
  • Platinum-cured silicone provides high purity and broad chemical compatibility.
  • Dimensions: top 12.5 mm x bottom 16.5 mm x height 20 mm (0.493 x 0.650 x 0.788 in).
  • Supplied 10 per pack for small-batch use and inventory control.
  • Intended for closure systems in single-use assemblies and laboratory glassware.
  • Datasheet and material compliance information available from the manufacturer.