Learn More
Accuflow Systems Saint-Gobain Silicone Stopper, 41 x 49 x 40 mm, 5/Pack
Catalog No. 502365840
Encompass_Preferred
Saint-Gobain Sani-Tech platinum-cured silicone stopper for laboratory and bioprocess applications. The stopper measures 41 x 49 x 40 mm and is supplied in packs of five. It provides an aseptic seal, resists cracking and spallation, shows no particle migration, and is fully autoclavable and sterilizable.
  • Ultra-pure, platinum-cured silicone.
  • Dimensions 41 x 49 x 40 mm.
  • Pack of 5 stoppers.
  • Fully autoclavable and sterilizable.
  • Resists cracking and spallation.
  • No particle migration; imparts no taste or odors.
  • Suitable for carboys, solution and media bottles, and laboratory apparatus.
Catalog No. 50-236-5840 Supplier Accuflow Systems Supplier No. D1069820
Saint-Gobain Sani-Tech platinum-cured silicone stopper for laboratory and bioprocess applications. The stopper measures 41 x 49 x 40 mm and is supplied in packs of five. It provides an aseptic seal, resists cracking and spallation, shows no particle migration, and is fully autoclavable and sterilizable.
  • Ultra-pure, platinum-cured silicone.
  • Dimensions 41 x 49 x 40 mm.
  • Pack of 5 stoppers.
  • Fully autoclavable and sterilizable.
  • Resists cracking and spallation.
  • No particle migration; imparts no taste or odors.
  • Suitable for carboys, solution and media bottles, and laboratory apparatus.