Learn More
Sorbent Technologies Inc Silica Gel for TLC, 60A, w/UV254, 5-15um, 500g
SDP
Catalog No. 501138713
Encompass_Preferred
Silica Gel for TLC with UV254 fluorescent indicator has a pore size of 60A, particle distribution of 5-15um, and a quantity of 500g. When combined with a binding agent, this silica gel can be used for the making of analytical TLC plates.
Catalog No. 50-113-8713 Supplier Sorbent Technologies Inc Supplier No. 1162005
Silica Gel for TLC with UV254 fluorescent indicator has a pore size of 60A, particle distribution of 5-15um, and a quantity of 500g. When combined with a binding agent, this silica gel can be used for the making of analytical TLC plates.