Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, Peroxide-Cured Silicone 1.52 mm ID; 6/PK
Catalog No. NC0385487
Encompass
Designed for optimal operation in your Ismatec pumps. It is resistant to ozone and UV light. It won't crack, swell or harden. Translucent yellow color. Temperature range: -50 to 74C (-58 to 165F).
Catalog No. NC0385487 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0762436
Designed for optimal operation in your Ismatec pumps. It is resistant to ozone and UV light. It won't crack, swell or harden. Translucent yellow color. Temperature range: -50 to 74C (-58 to 165F).