Learn More
Sorbent Technologies Inc Silica Gel, Standard Grade, 60A, 40-63um, 2.5kg
SDP
Catalog No. NC2962641
Encompass_Preferred
Silica Gel, Standard Grade has a controlled particle size distribution specific to flash chromatography applications. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 2.5kg. This silica gel provides good mechanical stability for longer silica life and is a good performance alternative for general separations in the lab. It is used for the packing of traditional flash chromatography columns.
Catalog No. NC2962641 Supplier Sorbent Technologies Inc Supplier No. 40930H2.5
Silica Gel, Standard Grade has a controlled particle size distribution specific to flash chromatography applications. The silica gel has a porosity of 60A, particle distribution of 40-63um (230x400mesh), and a quantity of 2.5kg. This silica gel provides good mechanical stability for longer silica life and is a good performance alternative for general separations in the lab. It is used for the packing of traditional flash chromatography columns.