Learn More
ATHENA ES SILAC- DMEM/F12
Catalog No. 5000158157
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.
Catalog No. 50-001-58157 Supplier ATHENA ES Supplier No. 0433
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.