Learn More
ATHENA ES SILAC- DMEM High Glucose with Phenol Red, powder
Catalog No. 5000158110
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids. Contains Penol Red.
Catalog No. 50-001-58110 Supplier ATHENA ES Supplier No. 0430P
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids. Contains Penol Red.