Learn More
ATHENA ES SILAC- DMEM Low Glucose, powder
Supplier:  ATHENA ES 0431P
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids. Contains Penol Red.
Catalog No. 50-001-58156