Learn More
ATHENA ES SILAC-IMDM
Catalog No. 5000158158
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.
Catalog No. 50-001-58158 Supplier ATHENA ES Supplier No. 0434
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.