Learn More
ATHENA ES SILAC - SFM Insect Medium
Catalog No. 5000158155
Encompass_Preferred
Special Order - SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.
Catalog No. 50-001-58155 Supplier ATHENA ES Supplier No. 0429
Special Order - SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.