Learn More
Electron Microscopy Sciences Silane Coated Slides, (aminoalkysilane) 72/bx
SDP
Catalog No. 5027990
Encompass_Preferred
Our premium glass slides with a frosted end are now available pre-coated. Pre-coated with Silane (aminoalkysilane), these slides improve the adhesion of tissues to the slide.
Catalog No. 50-279-90 Supplier Electron Microscopy Sciences Supplier No. 6341101
Our premium glass slides with a frosted end are now available pre-coated. Pre-coated with Silane (aminoalkysilane), these slides improve the adhesion of tissues to the slide.