Learn More
Electron Microscopy Sciences Silane Coated Slides, (aminoalkysilane) 144/bx
SDP
Catalog No. 5027987
Encompass_Preferred
Our premium glass slides with a frosted end are now available pre-coated. Pre-coated with Silane (aminoalkysilane), these slides improve the adhesion of tissues to the slide.
Catalog No. 50-279-87 Supplier Electron Microscopy Sciences Supplier No. 6341102
Our premium glass slides with a frosted end are now available pre-coated. Pre-coated with Silane (aminoalkysilane), these slides improve the adhesion of tissues to the slide.