Learn More
Analytical Sales & Services Silanized 9mm, 2mL Clear Screw Vials w/ 300µL Fused Insert and Cap w/ PTFE/Silicone Liners
Catalog No. 5000804459
Encompass
Advantage Silanized Unassembled Custom Vial Kit\n\nContains: 9mm, 2mL Clear Glass Screw Vials with 300µL Fused Glass Insert and 9mm Screw Cap with PTFE/Silicone Liners
Catalog No. 50-008-04459 Supplier Analytical Sales & Services Supplier No. 99042S
Advantage Silanized Unassembled Custom Vial Kit\n\nContains: 9mm, 2mL Clear Glass Screw Vials with 300µL Fused Glass Insert and 9mm Screw Cap with PTFE/Silicone Liners