Learn More
Alkali Scientific Silanized Glass Vials with PTFE/Silicone Lined Solid Closure, Pre assembled, Flat Bottom, White Writing Area with Graduations, Sterile, 100/Pk, 8mL (17mm), 15 425 Screw Thread, Clear
SDP
Catalog No. 5000663140
Encompass_Preferred
Save time and reduce waste costs during quantitative analysis or material storage with Alkali Scientific™ Silanized Glass Vials. These vials undergo a vapor phase deposition silanization process, ensuring surface deactivation to preserve the integrity of sensitive materials or extracts. By eliminating active sites on borosilicate glass surfaces, silanization and siliconization enhance sample stability and recovery. Silanized for efficiency – Minimizes waste costs and saves time during analysis and storage. Enhanced surface modification – The silylating agent reacts with active groups on the glass surface, reducing reactivity and preventing materials from adhering. Maximizes analyte recovery – Ideal for trace analysis by inhibiting sample loss due to surface interaction.
Catalog No. 50-006-63140 Supplier Alkali Scientific Supplier No. SV008
Save time and reduce waste costs during quantitative analysis or material storage with Alkali Scientific™ Silanized Glass Vials. These vials undergo a vapor phase deposition silanization process, ensuring surface deactivation to preserve the integrity of sensitive materials or extracts. By eliminating active sites on borosilicate glass surfaces, silanization and siliconization enhance sample stability and recovery. Silanized for efficiency – Minimizes waste costs and saves time during analysis and storage. Enhanced surface modification – The silylating agent reacts with active groups on the glass surface, reducing reactivity and preventing materials from adhering. Maximizes analyte recovery – Ideal for trace analysis by inhibiting sample loss due to surface interaction.