Learn More
Grainger CRC Silaramic Brake System Grease, Ceramic Lubricant, 5 oz
Catalog No. 502779117
Encompass_Preferred
Silaramic® Brake System Grease is a high-performance, pure silicone dry-film lubricant designed to protect brake systems under extreme operating conditions. Fortified with a high concentration of a unique ceramic lubricant, this grease effectively minimizes friction, wear, and corrosion. It is specifically formulated to remain stable at extreme temperatures, ensuring long-lasting performance for all brake components.
  • Extreme temperature range of -50°F to 3000°F
  • Protects against uneven pad wear, rust, and corrosion
  • Reduces brake noise and vibrations
  • Compatible with brake system rubber and plastics, including Nitrile
  • Will not wash out or melt in wet or dry conditions
  • Ideal for ceramic brake systems
Catalog No. 50-277-9117 Supplier Grainger Supplier No. 19MX66
Silaramic® Brake System Grease is a high-performance, pure silicone dry-film lubricant designed to protect brake systems under extreme operating conditions. Fortified with a high concentration of a unique ceramic lubricant, this grease effectively minimizes friction, wear, and corrosion. It is specifically formulated to remain stable at extreme temperatures, ensuring long-lasting performance for all brake components.
  • Extreme temperature range of -50°F to 3000°F
  • Protects against uneven pad wear, rust, and corrosion
  • Reduces brake noise and vibrations
  • Compatible with brake system rubber and plastics, including Nitrile
  • Will not wash out or melt in wet or dry conditions
  • Ideal for ceramic brake systems