Learn More
Hach Company Radiometer Analytical M291Ag-9 Metal Electrode (silver rod sensor, screw cap)
Supplier:  Hach Company E34M002
Encompass
The Radiometer M291Ag-9 Metal Electrode has a silver rod sensor and is recommended for silver halide titrations. The M291Ag-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for use with BNC-compatible meters/titrators.
Catalog No. 50-199-7600