Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, Platinum-Cured Silicone, I/P 26; 200 ft
Catalog No. NC0506511
Encompass
Reduce the risk of particulate entrapment thanks to the smooth inner surface. Genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.
Catalog No. NC0506511 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640326/DEL
Reduce the risk of particulate entrapment thanks to the smooth inner surface. Genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.