Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, Platinum-Cured Silicone, I/P 82; 100 ft
Catalog No. NC9950789
Encompass
Reduce the risk of particulate entrapment thanks to the smooth inner surface. Genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.
Catalog No. NC9950789 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640382
Reduce the risk of particulate entrapment thanks to the smooth inner surface. Genuine Masterflex tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility. Made by a global market leader in range of formulations for most extensive chemical compatibility.