Learn More
Selleck Chemical LLC Sildenafil-100mg
SDP
Catalog No. 502389678
Encompass_Preferred
Sildenafil (Revatio, UK-92480, Viagra) is a Phosphodiesterase 5 Inhibitor with IC50 of 5.22 nM.
Catalog No. 50-238-9678 Supplier Selleck Chemical LLC Supplier No. S4684100MG
Sildenafil (Revatio, UK-92480, Viagra) is a Phosphodiesterase 5 Inhibitor with IC50 of 5.22 nM.