Learn More
Medchemexpress LLC Sildenafil | 139755-83-2 | C22H30N6O4S | 1 G
SDP
Catalog No. 5000375356
Encompass_Preferred
Sildenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It is intended for research use only.
  • Potent PDE5 inhibitor
  • IC50 of 5.22 nM against PDE5
  • Potentiates ERK1/ERK2 phosphorylation
  • Increases cell proliferation
  • Increases intracavernosal pressure and ICP/blood pressure ratio in dog models
  • Decreases the number of TL+-cells
  • Reduces microglia/macrophages in pMCAo models
  • Decreases flap necrosis in preclinical animal models
Catalog No. 50-003-75356 Supplier Medchemexpress LLC Supplier No. HY150251G
Sildenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It is intended for research use only.
  • Potent PDE5 inhibitor
  • IC50 of 5.22 nM against PDE5
  • Potentiates ERK1/ERK2 phosphorylation
  • Increases cell proliferation
  • Increases intracavernosal pressure and ICP/blood pressure ratio in dog models
  • Decreases the number of TL+-cells
  • Reduces microglia/macrophages in pMCAo models
  • Decreases flap necrosis in preclinical animal models