Learn More
Medchemexpress LLC Sildenafil | 139755-83-2 | C22H30N6O4S | 200 MG
SDP
Catalog No. 5000375536
Encompass_Preferred
Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It demonstrates various cellular and in vivo activities, making it suitable for a range of research applications. This product is for research use only.
  • Potent PDE5 inhibitor with an IC50 of 5.22 nM
  • Inhibits human recombinant PDE5A1
  • Potentiates ERK1/ERK2 phosphorylation
  • Increases cell proliferation in S phase
  • Significantly increases intracavernosal pressure (ICP) in dog models
  • Reduces number of TL+-cells and microglia/macrophages in vivo
  • Decreases flap necrosis in animal models by enhancing growth factor secretion
Catalog No. 50-003-75536 Supplier Medchemexpress LLC Supplier No. HY15025200MG
Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It demonstrates various cellular and in vivo activities, making it suitable for a range of research applications. This product is for research use only.
  • Potent PDE5 inhibitor with an IC50 of 5.22 nM
  • Inhibits human recombinant PDE5A1
  • Potentiates ERK1/ERK2 phosphorylation
  • Increases cell proliferation in S phase
  • Significantly increases intracavernosal pressure (ICP) in dog models
  • Reduces number of TL+-cells and microglia/macrophages in vivo
  • Decreases flap necrosis in animal models by enhancing growth factor secretion