Learn More
Medchemexpress LLC Sildenafil | 139755-83-2 | C22H30N6O4S | 50 MG
SDP
Catalog No. 502029079
Encompass_Preferred
Sildenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. This compound is intended for research use only and is not sold to patients.
  • Potent phosphodiesterase type 5 (PDE5) inhibitor
  • Cited in numerous scientific publications
  • Potentiates phosphorylation of ERK1/ERK2 and increases cell proliferation in vitro
  • Significantly increases intracavernosal pressure in a dog model of erection
  • Reduces microglia/macrophages in a pMCAo mouse model
  • Decreases flap necrosis in preclinical animal models by increasing growth factors
  • Under investigation in clinical trials for various conditions including Heart Failure and Erectile Dysfunction
Catalog No. 50-202-9079 Supplier Medchemexpress LLC Supplier No. HY1502550MG
Sildenafil is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. This compound is intended for research use only and is not sold to patients.
  • Potent phosphodiesterase type 5 (PDE5) inhibitor
  • Cited in numerous scientific publications
  • Potentiates phosphorylation of ERK1/ERK2 and increases cell proliferation in vitro
  • Significantly increases intracavernosal pressure in a dog model of erection
  • Reduces microglia/macrophages in a pMCAo mouse model
  • Decreases flap necrosis in preclinical animal models by increasing growth factors
  • Under investigation in clinical trials for various conditions including Heart Failure and Erectile Dysfunction