Learn More
Cayman Chemical SIldenafIl ChLRosulfonyl 500mg
SDP
Catalog No. 502910220
Encompass_Preferred
An intermediate in the synthesis of sildenafil; a potential impurity found in commercial preparations of sildenafil
Catalog No. 50-291-0220 Supplier Cayman Chemical Supplier No. 34760500 MG
An intermediate in the synthesis of sildenafil; a potential impurity found in commercial preparations of sildenafil