Learn More
Medchemexpress LLC HY-15025A 100mg , Sildenafil (citrate) CAS:171599-83-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15025A100MG
Encompass_Preferred
HY-15025A 100mg Sildenafil (citrate) CAS: 171599-83-0 Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with IC50 of 5.22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0888