Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sildenafil citrate

Catalog No. AC463250010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463250010 Supplier Thermo Scientific Chemicals Supplier No. 463250010

CAS: 171599-83-0 | C28H38N6O11S | 666.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171599-83-0
Molecular Formula C28H38N6O11S
Molecular Weight (g/mol) 666.70
InChI Key DEIYFTQMQPDXOT-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

Specifications

Melting Point 182.0°C to 196.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: very slightly soluble. Other solubilities: insoluble in ether & acetonitrile
Formula Weight 666.7
Chemical Name or Material Sildenafil citrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Causes damage to organs.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.