Learn More
Medchemexpress LLC Sildenafil citrate | 171599-83-0 | MFCD09026931 | C22H30N6O4S ·C6H8O7 | 500 MG
SDP
Catalog No. 5000375326
Encompass_Preferred
Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It has demonstrated efficacy in potentiating the phosphorylation of ERK1/ERK2, promoting cell proliferation, and enhancing the upregulation of ERK1/ERK2 phosphorylation induced by serotonin in in vitro studies. In vivo, Sildenafil citrate has been shown to significantly increase intracavernosal pressure (ICP) and ICP/blood pressure (BP) in dog models of erection, and it has been reported to decrease flap necrosis in preclinical animal models.
  • Potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM
  • Potentiates ERK1/ERK2 phosphorylation in vitro
  • Promotes cell proliferation and enhances ERK1/ERK2 phosphorylation induced by serotonin
  • Increases intracavernosal pressure (ICP) and ICP/BP in dog models of erection
  • Reported to decrease flap necrosis in preclinical animal models
Catalog No. 50-003-75326 Supplier Medchemexpress LLC Supplier No. HY15025A500MG
Sildenafil citrate is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. It has demonstrated efficacy in potentiating the phosphorylation of ERK1/ERK2, promoting cell proliferation, and enhancing the upregulation of ERK1/ERK2 phosphorylation induced by serotonin in in vitro studies. In vivo, Sildenafil citrate has been shown to significantly increase intracavernosal pressure (ICP) and ICP/blood pressure (BP) in dog models of erection, and it has been reported to decrease flap necrosis in preclinical animal models.
  • Potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM
  • Potentiates ERK1/ERK2 phosphorylation in vitro
  • Promotes cell proliferation and enhances ERK1/ERK2 phosphorylation induced by serotonin
  • Increases intracavernosal pressure (ICP) and ICP/BP in dog models of erection
  • Reported to decrease flap necrosis in preclinical animal models