Learn More
Apexbio Technology LLC Sildenafil Citrate, 25mg. Cas: 171599-83-0 MFCD: MFCD02102122. Treat erectile dysfunction and PAH
SDP
Supplier:  Apexbio Technology LLC A432125
Encompass_Preferred
Sildenafil Citrate is a potent inhibitor of cGMP-specific phosphodiesterase including PDE5 with IC50 values of 3.6 nM.[1] PDE5 (cGMP-specific phosphodiesterase type 5) is an enzyme from the phosphodiesterase class. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4923
 Restricted Item