Learn More
Selleck Chemical LLC Sildenafil Citrate S1431-1g
SDP
Catalog No. 503110066
Encompass_Preferred
Sildenafil Citrate a selective inhibitor of cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5) is a well-tolerated and highly effective treatment for erectile dysfunction
Catalog No. 50-311-0066 Supplier Selleck Chemical LLC Supplier No. S14311G
Sildenafil Citrate a selective inhibitor of cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5) is a well-tolerated and highly effective treatment for erectile dysfunction