Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sildenafil Citrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606194 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-194 100 g Amber Glass Bottle
18-606-196 500 g Amber Glass Bottle
18-606-195 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-194 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S3092100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S3092, 171599-83-0, C22H30N6O4S·C6H8O7

Spectrum™ Chemical Sildenafil Citrate, USP

Chemical Identifiers

CAS 171599-83-0
Molecular Formula C28H38N6O11S
Molecular Weight (g/mol) 666.70
InChI Key DEIYFTQMQPDXOT-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.