Learn More
Selleck Chemical LLC Sildenafil Citrate 25mg 171599-83-0
SDP
Catalog No. 506318
Encompass_Preferred
Sildenafil Citrate, a selective inhibitor of cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5), is a well-tolerated and highly effective treatment for erectile dysfunction. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-631-8 Supplier Selleck Chemical LLC Supplier No. S143125MG
Sildenafil Citrate, a selective inhibitor of cyclic guanosine monophosphate (cGMP)-specific phosphodiesterase type 5 (PDE5), is a well-tolerated and highly effective treatment for erectile dysfunction. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.