Learn More
Cayman Chemical SIldenafIl-d3 1mg
SDP
Catalog No. 502966121
Encompass_Preferred
An internal standard for the quantification of sildenafil by GC- or LC-MS
Catalog No. 50-296-6121 Supplier Cayman Chemical Supplier No. 182611 MG
An internal standard for the quantification of sildenafil by GC- or LC-MS