Learn More
Cayman Chemical SIldenafIl-d3 500ug
SDP
Catalog No. 502966754
Encompass_Preferred
An internal standard for the quantification of sildenafil by GC- or LC-MS
Catalog No. 50-296-6754 Supplier Cayman Chemical Supplier No. 18261500 UG
An internal standard for the quantification of sildenafil by GC- or LC-MS