Learn More
Medchemexpress LLC Sildenafil-d3N-1 | 1126745-87-6 | C22H27D3N6O4S | 5 MG
SDP
Catalog No. 5000375921
Encompass_Preferred
Sildenafil-d3-1 is a deuterium-labeled Sildenafil-d3. It is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. This compound has a molecular weight of 477.59, appears as a white to off-white solid, and has a purity of 99.47%. It is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-75921 Supplier Medchemexpress LLC Supplier No. HY15025S25MG
Sildenafil-d3-1 is a deuterium-labeled Sildenafil-d3. It is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. This compound has a molecular weight of 477.59, appears as a white to off-white solid, and has a purity of 99.47%. It is intended for research use only and not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS