Learn More
Medchemexpress LLC Sildenafil-d3n-1 | 1126745-87-6 | MFCD09841266 | 99.5% | 477.59 g/mol | C22H27D3N6O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15025S210MG
Encompass_Preferred
Sildenafil-d3-1 is a deuterium-labeled analogue of sildenafil supplied for research and analytical use. It serves as a labeled reference standard and analytical internal standard for quantitative assays and pharmacokinetic studies, and is provided with supporting documentation for laboratory handling and verification.
  • Deuterium-labeled analogue for use as an internal standard in LC-MS/MS and GC-MS assays.
  • High purity (99.47%) for reproducible analytical performance.
  • Molecular formula C22H27D3N6O4S and molecular weight 477.59 g/mol.
  • Suitable for ADME, pharmacokinetic, and stability studies.
  • Supplied as a solid with certificate of analysis, safety data sheet, and datasheet.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-86705