Learn More
Medchemexpress LLC Sildenafil-d8 | 951385-68-5 | C22H22D8N6O4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15025S11MG
Encompass_Preferred
Sildenafil-d8 is a deuterium-labeled form of Sildenafil, which is recognized as a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitative tracing during the drug development process. Deuteration can potentially influence the pharmacokinetic and metabolic characteristics of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-75526