Learn More
Selleck Chemical LLC Sildenafil Mesylate S4683-500mg
SDP
Catalog No. 503109524
Encompass_Preferred
Sildenafil Mesylate(UK-92480 Mesylate) is a mesylate form of Sildenafil an inhibitor of Phosphodiesterase 5
Catalog No. 50-310-9524 Supplier Selleck Chemical LLC Supplier No. S4683500MG
Sildenafil Mesylate(UK-92480 Mesylate) is a mesylate form of Sildenafil an inhibitor of Phosphodiesterase 5