Learn More
Lonza Walkersville Silensomes CYP Phenotyp Kit
Catalog No. 5000496211
Encompass
Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for one specific CYP using mechanism based inhibitors (MBI) Contents of the CYP Phenotyping Kit include 7 individual Silensomes HLM inhibited for CYP2C9 CYP1A2 CYP2B6 CYP2A6 CYP2D6 CYP2C8 and CYP3A4 P450s plus the Silensomes Master Control Format Each specific CYP Silensomes HLM is supplied in 150 L as 20mg/mL microsomal protein with matched Control Silensome HLM (batch dependent) that was treated under the exact same conditions without inhibitor
Catalog No. 50-004-96211 Supplier Lonza Walkersville Supplier No. SILMCPNL
Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for one specific CYP using mechanism based inhibitors (MBI) Contents of the CYP Phenotyping Kit include 7 individual Silensomes HLM inhibited for CYP2C9 CYP1A2 CYP2B6 CYP2A6 CYP2D6 CYP2C8 and CYP3A4 P450s plus the Silensomes Master Control Format Each specific CYP Silensomes HLM is supplied in 150 L as 20mg/mL microsomal protein with matched Control Silensome HLM (batch dependent) that was treated under the exact same conditions without inhibitor