Learn More
Lonza Walkersville Silensomes CYP1A2 Kit w/Contr
Catalog No. 5000496242
Encompass
CYP1A2 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP1A2 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM
Catalog No. 50-004-96242 Supplier Lonza Walkersville Supplier No. SIL210K
CYP1A2 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP1A2 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM