Learn More
Lonza Walkersville Silensomes CYP2D6 Kit w/Contr
Catalog No. 5000496210
Encompass
CYP2D6 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP2D6 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM
Catalog No. 50-004-96210 Supplier Lonza Walkersville Supplier No. SIL240K
CYP2D6 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP2D6 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM