Learn More
Lonza Walkersville Silensomes CYP3A4 Kit w/Contr
Catalog No. 5000496322
Encompass
CYP3A4 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP3A4 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM
Catalog No. 50-004-96322 Supplier Lonza Walkersville Supplier No. SIL200K
CYP3A4 Silensomes are validated human pooled liver microsomes (HLM) chemically and irreversibly inactivated for CYP3A4 using a mechanism based inhibitor (MBI) Included is a production-matched control HLM